Kimberly-Clark Professional™ Kimtech™ N95 Pouch Respirator
N95 pouch respirator is NIOSH-Approved and provides a minimum 95% filtration efficiency of 0.3 micron particles.
Supplier:  Kimberly-Clark Professional™ 53358
 View more versions of this product
Catalog No. 19-005-123
Explore available promotions
Alert:
Not for medical use. Not to be used by healthcare professionals or in healthcare settings.
Description
Description
  • NIOSH-Approved, provides a minimum 95% filtration efficiency of 0.3 micron particles
  • Guards against the risks posed by airborne particles and contaminants
  • Pouch-style offers the wearer a large breathing chamber to maximize comfort
  • Equipped with two comfort fit headbands and a bendable nosepiece to ensure a consistent seal
  • Not intended for the healthcare market and/or healthcare personnel
  • 300 Kimtech™ N95 Pouch Respirators (6 bags of 50)
Specifications
Specifications
N - Not Resistant to Oil
White
Single-use
General Purpose Particulate Respirator
No
53358
6/Bag, 50 Bags/Cs.
Two Comfort Fit Headbands
NIOSH 42 CFR 84, N95
Pouch/Duckbill
N95
One Size
95%
Adjustable Nose Clip
Non-sterile
No