missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly Clark Kimtech™ Polaris™ Nitrile Exam Gloves
Kimtech™ is your partner in driving progress. Tested against some of the harshest chemicals used in your work environment, Kimtech™ Polaris™ Exam Nitrile Gloves meet and exceed the industry standards for exam gloves.
$33.77 - $33.77
Specifications
Chlorinated | No |
---|---|
Accelerator Free | No |
Applications | Laboratory |
AQL | 0.65 |
Color | Dark Magenta |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-103-196A
|
Kimberly Clark
62770 |
X-Small |
Pack of 100 Each for $33.77
Case of 10 Pack for $322.60
|
|
|||||
17-103-196B
|
Kimberly Clark
62771 |
Small |
Pack of 100 Each for $33.77
Case of 10 Pack for $322.60
|
|
|||||
17-103-196C
|
Kimberly Clark
62772 |
Medium |
Pack of 100 Each for $33.77
Case of 10 Pack for $322.60
|
|
|||||
17-103-196D
|
Kimberly Clark
62773 |
Large |
Pack of 100 Each for $33.77
Case of 10 Pack for $322.60
|
|
|||||
17-103-196E
|
Kimberly Clark
62774 |
X-Large |
Pack of 90 Each for $33.77
Case of 10 Pack for $322.60
|
|
|||||
Description
- As part of Kimberly Clark's 150-year-old legacy of innovation and partnership with the scientific community, we're proud to present our next step into the future— Kimtech™ Polaris™ Nitrile Gloves.
- The proprietary high-quality nitrile formulation delivers the highest level of protection in the Kimtech™ exam glove portfolio and has been tested for use with chemotherapy drugs and fentanyl.
- With an average tensile strength that is more than double the industry standard, Kimtech™ Polaris™ provides the strength and comfort needed for performance and productivity; it's even certified by US Ergonomics.
- Eligible for recycling through The RightCycle™ Program by Kimberly-Clark Professional™, 100% of your non-hazardous Kimtech™ Polaris™ Nitrile Exam Gloves can be diverted from the landfill.
- With protection, durability, and comfort you can count on, these gloves are built for science, made for you
- Ambidextrous
Specifications
No | |
Laboratory | |
Dark Magenta | |
Fingertip Textured | |
0.15 mm | |
Nitrile | |
0.15 mm | |
Nitrile Examination Glove | |
5.91 mil | |
Yes | |
9.5 in. |
No | |
0.65 | |
Single-use | |
5.91 mil | |
Yes | |
5.91 mil | |
Powder Free | |
Non-sterile | |
0.15 mm | |
Beaded | |
24.13 cm |