Kimberly-Clark Professional™ Kimtech™ Prep Wipers
Designed for use in WetTask™ Refillable Wet Wiping System
Supplier:  Kimberly-Clark Professional™ 06411
 View more versions of this product
Catalog No. 19-130-2378
Explore available promotions
Description
Description
  • System is an ideal alternative to open buckets and rags; reduces exposure to chemical vapors and splashes
  • Enclosed system helps avoid contamination of wipers and cleaning solution
  • Maintains concentration of actives released at near target levels over a prolonged period of use
Specifications
Specifications
Critical Task Wipe
31.75 cm
30.48 cm
White
12.5 x 12 in. (31.75 x 30.48 cm)
Bleach, quaternary amine disinfectants and sanitizer
90 Sheets/Roll
12.5 in.
12.5 in.
Meltblown
Single-use
Roll
WetTask™ refillable wet wiping system
Product Suggestions
Product Suggestions