missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ Kimtech™ A8 Cleanroom Coveralls

Cleanroom coveralls are designed to enhance comfort and reduce heat stress in ISO 7 and 8 environments.

$180.24 - $220.56

Specifications

Product Type Cleanroom Coveralls
Material SMS
Color White
Disposable Single-use
Cuff Style Thumb Loop
View More Specs

Products 7
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-169-222A
View Documents
Kimberly-Clark Professional™
47951
Small
Case of 25 Each for $180.24
 
19-169-222B
View Documents
Kimberly-Clark Professional™
47952
Medium
Case of 25 Each for $184.12
 
19-169-222C
View Documents
Kimberly-Clark Professional™
47953
Large
Case of 25 Each for $189.16
 
19-169-222D
View Documents
Kimberly-Clark Professional™
47954
X-Large
Case of 25 Each for $191.88
 
19-169-222E
View Documents
Kimberly-Clark Professional™
47955
2X-Large
Case of 25 Each for $202.73
 
19-169-222F
View Documents
Kimberly-Clark Professional™
47956
3X-Large
Case of 25 Each for $216.29
 
19-169-222G
View Documents
Kimberly-Clark Professional™
47957
4X-Large
Case of 25 Each for $220.56
 
Description

Description

Kimtech™ A8 Cleanroom Coveralls are made with a lightweight, innovative fabric that moves with the worker, and feature built-in thumb loops to protect your people and processes, and a high neck collar to control contamination risk. These white coveralls also have elastic wrists, ankles and waist.

They are designed for protection against dust, chemicals and other contaminants and have a BFE of 94%. The white coveralls are designed for applications including ISO 7/8 cleanrooms, Grade C/D, non-sterile pharmaceutical manufacturing, bio-medical research, pharmaceutical compounding and biotechnology.

Plus, these protective suits are recyclable through the RightCycle Program by Kimberly-Clark Professional.

  • 25 Kimtech™ A8 Coveralls/Case
  • Helps protect against dust, chemicals and other contaminants; BFE of 94%; ISO 7/8 cleanrooms
  • Low lint for contamination control
  • Recyclable through the Kimberly-Clark Professional™ RightCycle Program, which turns used lab coats into raw materials for eco-friendly consumer products
Specifications

Specifications

Cleanroom Coveralls
White
Thumb Loop
Non-sterile
Standing
Kimtech™ A8
Double Bagged
SMS
Single-use
Collared
Anti-static
PPE Cat III directive 89/686/EEC, Type 6 limited chemical splash protection; Type 5 particle protection
Pharmaceutical, Packing, Distribution, Production, Biotechnology
Class 10,000-100,000 (ISO 7-8)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'