missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ Kimtech™ Pure M5 Pleat-Style Cleanroom Face Masks

Three-layer, pleat-style face masks for use in ISO Class 5 and higher cleanroom environments. Available with earloops or soft ties.

$120.40 - $199.14

Specifications

Product Type Cleanroom Face Mask
Color Blue
Material Polyester
Size Standard
Sterility Non-sterile
View More Specs

Products 2
Catalog Number Mfr. No. Fastening Type Price Quantity  
Catalog Number Mfr. No. Fastening Type Price Quantity  
19-062279
View Documents
Kimberly-Clark Professional™
62692
Earloops
Case of 500 Each for $199.14
 
29-636-0582
View Documents
Kimberly-Clark Professional™
62742
Ties
Case of 300 Each for $120.40
 
Description

Description

  • Three-layer design
  • Made with polyester and cellulose
  • Pleat-style masks available with earloops or soft ties
  • Fully enclosed bendable nosepiece
  • Popular for biotech, medical device, electronics and telecommunication environments
  • Ultrasonically bonded ties are durable
  • Double bagged and recommended for ISO Class 5 and higher cleanroom environments
Specifications

Specifications

Cleanroom Face Mask
Polyester
Non-sterile
Class 100 (ISO 5)
Single-use
Biotech, Medical Devices, Electronics, Telecommunications
M5
Blue
Standard
Pleated
3-ply
99%
Double-bagged
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'