missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ KingFisher Apex Purification Systems
The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source.
Supplier: Thermo Scientific™ A46941
Description
The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source. It moves magnetic beads, not liquids, through the main steps of sample preparation: bind, wash, and elute. This process provides excellent quality, reproducible results, and reduces hands-on time for sample processing.
- User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
- Provides versatile and high-speed purification with reproducible results
- Easy to use interface with touch screen empowering the user to control all details of the application and purification process
- BindIx PC Software for added convenience
- Extensive selection of validated protocols for different type of applications
- Cloud library for quick protocol retrieval
Specifications
Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate | |
10 to 80 μL | |
96 Samples | |
>98% with 2.8 μm particle size | |
Touch screen (10 in.) | |
+4°C above ambient temperature up to +100°C | |
ca. >1 μm | |
96 | |
Up to 23 hr. 59 min. | |
96 PCR plate (semi/skirted) | |
56 cm | |
46 cm | |
56 kg | |
79 cm | |
1 year manufacturer warranty and 1 year of field service with PM |
PCR/qPCR, Sequencing, Mass Spec | |
8 | |
KingFisher Apex with 96 PCR head, Extended Warranty Package | |
Down to +4°C | |
Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | |
96 | |
System + SmartStart + 1 yr ext warranty | |
96 in one frame, interchangeable | |
2 Pc. | |
22 in. | |
18 in. | |
123 lb. | |
31 in. | |
1 day at customer site |