Thermo Scientific™ KingFisher™ Flex 96 heating block DFS Item
The KingFisher™ Flex 96 KF Heating Block is compatible with the KingFisher™ 96 KF Microplate (200 μL) and the MagMAX™ Express-96 Standard Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 96 Deep-Well Head.
Supplier:  Thermo Scientific™ 24075420
Catalog No. 22387018
Explore available promotions
Description
Description
The KingFisher™ Flex 96 KF Heating Block is compatible with the KingFisher™ 96 KF Microplate (200 μL) and the MagMAX™ Express-96 Standard Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 96 Deep-Well Head. This heating block is also compatible with the KingFisher™ Flex Microtiter Deep Well 96 Plate and the MagMAX™ Express-96 Deep Well plates.
Specifications
Specifications
96-well Plate
KingFisher™ Flex
Each
96 Samples
KingFisher™
Room Temperature
Videos
SDS
Documents
Documents
Promotions
Promotions
For research use only. Not for use in diagnostic procedures.