Learn More
Thermo Scientific™ KingFisher™ Flex Purification System
Prepare samples efficiently from a variety of materials with Thermo Scientific KingFisher Flex Purification Systems, which offer highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source.
Supplier: Thermo Scientific™ A39952
Description
Prepare samples efficiently from a variety of materials with Thermo Scientific KingFisher Flex Purification Systems, which offer highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality.
Find a flexible solution for different types of sample processing needs. With 24 configurations, customers can raise the processing volume up to 5mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.
• Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
• High throughput system processes up to 96 samples in less than 20 minutes.
• Expands sample volume by 10-fold with the 24-well format.
• Open and flexible system lets the customer use anymagnetic particle-based kit tomeet the application demands.
• Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation andmodification.
• Ready-made protocols for different type of applications are available.
Recommended for:
• Genomics and proteomics
• Target identification
• Veterinary assays
• Biomarker discovery
• Quality control
• Forensics
Specifications
20 to 200 μL | |
96-well Plate | |
KingFisher 96 Plate, Extended Warranty Package | |
START / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / Four cursor keys / LCD | |
From +5°C above ambient temperature to +115°C | |
ca. >1 μm | |
24 or 96-well plates; KingFisher 24 Deep Well plate (200 to 5000 μL); Microtiter 96 Deep Well plate (50 to 1000 μμL); KingFisher 96 plate (20 to 200 μL);PCR plate (20 to 100 μL) | |
60 cm | |
38 cm | |
28 kg | |
68 cm | |
1 year manufacturer warranty and 1 year of field service with PM |
24 or 96 | |
96 Samples | |
> 95% | |
Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | |
96 in one frame; Interchangeable magnet heads | |
System + SmartStart + 1 yr ext warranty | |
23.6 in. | |
15 in. | |
62 lb. | |
26.8 in. | |
1 day at customer site |
For Laboratory Use