Thermo Scientific™ KingFisher™ Presto Purification System
Use the Thermo Scientific™ KingFisher™ Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories.
Supplier: Thermo Scientific™ 24078840
Description
Features:
- Purify 96 or 24 samples at a time
- Volume range of 50-5000μL
- Minimized size for easy fitting to automation systems
- Heating capability for application requiring elevated temperatures
- Easy connection to several liquid handling instruments
- Flexible system for wide variety of applications
Specifications
200 to 5000 μL | |
KingFisher Presto 24 Deep Well head | |
KingFisher™ Presto Purification System | |
24 |