BUCHI Kjeldahl Sample Tubes
Sample tubes for standard Kjeldahl are suitable for most applications.
Supplier: BUCHI 037377
Description
- Precision joint: tightness, avoid fume leaks
- Constricted condensation zone; no splashing
- Wider diameter; reduced foaming
- Thicker glass wall, longer lifetime
Specifications
Standard Kjeldahl | |
Sample Tube | |
300 mL | |
4/Pk. |