missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A40 Coveralls, Elastic Wrists and Ankles
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
$172.28 - $220.34
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | Microporous Film Laminate |
Color | White |
Disposable | Limited-use |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-120-2399H
|
Kimberly-Clark Professional™
30904 |
5X-Large |
Case of 25 Each for $220.34
|
|
|||||
17000254
|
Kimberly-Clark Professional™
30905 |
Small |
Case of 25 Each for $172.28
|
|
|||||
19-120-2399B
|
Kimberly-Clark Professional™
44312 |
Medium |
Case of 25 Each for $195.47
|
|
|||||
19-120-2399C
|
Kimberly-Clark Professional™
44313 |
Large |
Case of 25 Each for $195.47
|
|
|||||
19-120-2399D
|
Kimberly-Clark Professional™
44314 |
X-Large |
Case of 25 Each for $174.84
|
|
|||||
19-120-2399E
|
Kimberly-Clark Professional™
KC44315 |
2X-Large |
Case of 25 Each for $195.47
|
|
|||||
19-120-2399F
|
Kimberly-Clark Professional™
44316 |
3X-Large |
Case of 25 Each for $206.92
|
|
|||||
19-120-2399G
|
Kimberly-Clark Professional™
44317 |
4X-Large |
Case of 25 Each for $212.08
|
|
|||||
Description
Protective coveralls are made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
- Breathable, microporous film laminate fabric.
- Barrier protection against hazardous particulates and aerosols and against low-hazard liquid splash.
- ASTM F1670 for blood penetration and ASTM F1671 for bloodborne pathogens.
- ASTM F903 Penetration Testing provides passing results for isocyanate paint (40%) and Potassium Chromate (20%).
- Anti-stat according to NFPA 99.
- Features a zippered front and come in a variety of sizes.
- Eligible for recycling via RightCycle by Kimberly-Clark Professional™ when used in non-hazardous applications as classified by federal, state, and local regulations.
- Elastic wrists and ankles.
- Human-centric Reflex™ design offers comfortable fit and freedom of movement.
Specifications
General Purpose Coveralls | |
Microporous Film Laminate | |
Limited-use | |
Elastic | |
Unhooded | |
Non-sterile | |
ASTM F1670, ASTM F1671, ASTM F903, NFPA 99 | |
Fiberglass Manufacturing, Liquid Handling, Utilities, Pressure Washing, Tank Cleaning, Maintenance and General Manufacturing |
Unisex | |
White | |
Elastic | |
Collared | |
Front Zipper | |
Particle, Liquid | |
KleenGuard™ A40 | |
Elastic Wrists and Ankles |