missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A40 Reflex™ Liquid and Particle Protection Coveralls
Coveralls feature a respirator fit hood, zipper front, elastic wrists and ankles. Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
$215.13 - $222.11
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | Microporous Film Laminate |
Color | White |
Disposable | Limited-use |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-005-124
|
Kimberly-Clark Professional™
47995 |
Medium |
Case of 25 Each for $215.13
|
|
|||||
19-005-126
|
Kimberly-Clark Professional™
47997 |
X-Large |
Case of 25 Each for $218.23
|
|
|||||
19-005-127
|
Kimberly-Clark Professional™
47998 |
2X-Large |
Case of 25 Each for $220.56
|
|
|||||
19-005-128
|
Kimberly-Clark Professional™
47999 |
3X-Large |
Case of 25 Each for $222.11
|
|
|||||
Description
- Feature respirator fit hood, zipper front, elastic wrists and ankles
- Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates
- Pass NFPA 99 for antistatic materials
- Pass ASTM F1670 testing for penetration of blood and bodily fluids
- Reflex sizing for a more comfortable fit and less rip outs
- Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing
Specifications
General Purpose Coveralls | |
Microporous Film Laminate | |
Limited-use | |
Elastic | |
Elastic, Respirator Fit | |
Serged | |
Liquid, Particle, Anti-static, Chemicals, Bloodborne Pathogens | |
KleenGuard™ A40 Reflex™ | |
Aviation, Pharmaceutical Manufacturing, General Manufacturing, Liquid Handling |
Unisex | |
White | |
Elastic | |
Hooded | |
Front Zipper | |
Non-sterile | |
NFPA 99, ASTM F1670 | |
None |