Kimberly-Clark Professional™ KleenGuard™ Nemesis™ Eyewear
KleenGuard™ Nemesis™ Safety Eyewear is stylish, durable and have a lightweight construction.
$70.34 - $84.15
Specifications
Certifications/Compliance Meets ANSI/ISEA Z87.1+ standards.
Product Type Safety Glasses
Frame Color Black
Includes Neck Cord
Material Plastic
View More Specs
Products 2
Catalog Number Mfr. No. Lens Tint Lens Coating Price Quantity  
Catalog Number Mfr. No. Lens Tint Lens Coating Price Quantity  
19-150-0471
Accessories View Documents
Kimberly-Clark Professional™
25676
Clear Uncoated
Case of 12 Each for $70.34
19-014-380
View Documents
Kimberly-Clark Professional™
25679
Clear UV-treated, Anti-fog
Case of 12 Each for $84.15
Description
Description
KleenGuard™ Nemesis™ Safety Eyewear's stylish good looks and durable, lightweight construction are just some of the reasons why millions of people wear them for protection both at work and at home.
Nemesis™ Safety Glasses have polycarbonate lenses that provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss. The wraparound frame has a base curve of 8 that extends the range of peripheral protection, as well as soft touch temples for comfort.
The Nemesis™ safety glasses have a patented nose piece designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose – all for long-term comfort.
Each pair of KleenGuard™ Nemesis™ safety glasses come with a neck cord for securing them, so that the glasses are ready to protect at a moment's notice. The safety eyewear is manufactured to comply with the ANSI/ISEA Z87.1+ standard for personal eye protection.
Both men and women will find these stylish glasses to be comfortable and functional for construction, manufacturing, gun shooting / shooting range, and a host of other industries.
  • Ideal for indoor use
  • Feature a sporty, flexible, lightweight design that your employees will be happy to wear; they have a stylish, sporty look and feel
  • Meets ANSI Z87.1+ standards for personal eye protection
  • Polycarbonate lenses provide 99.9% UVA/UVB/UVC
  • Patented FLEX-DRY designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose – all for long-term comfort
Compliance
Meet ANSI/ISEA Z87.1+ standards.
Specifications
Specifications
Meets ANSI/ISEA Z87.1+ standards.
Black
Plastic
Plastic
Safety Glasses
Neck Cord
Reusable
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'