Kimberly-Clark Professional™ KleenGuard™ V60 Dual Readers Safety Eyewear
When you need vision assistance and eye protection, you can trust KleenGuard Dual Readers. They feature integrated diopters (+1.5 strength) on both the top and the bottom for vision help when looking both up and down.
Supplier:  Kimberly-Clark Professional™ 20389
 View more versions of this product
Catalog No. 19-169-912
Explore available promotions
Description
Description
  • Ideal for when workers need vision assistance and eye protection
  • Stylish, nylon frames feature a soft, self-adjusting rubber nose pad for comfort
  • Clear lenses allow maximum visible light transmission (ideal for indoor use) and are hard coated for scratch resistance and longer life
  • Meet ANSI Z87.1+ standards for high impact
  • Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
  • Come in three different strengths (+1.5, +2.0 and +2.5)
  • 7 base curve, which helps provide for peripheral vision and protection and feature an hard coating to prevent lens damage
Specifications
Specifications
ANSI Z87.1
Dual Readers Safety Eyewear
Black
UV400
Clear
For performing tasks that require up and down near vision or working upside down or overhead
+2.50