missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Knife Switch

Knife Switch on a durable plastic base. Rugged construction that stands up to tough student use. Poles and knives are made of brass for good electrical connections.

Supplier:  Eisco™ PH1000A

 View more versions of this product

Catalog No. S23687

Explore available promotions

Description

Description

  • Durable plastic base knife switch
  • Brass poles and knives for good electrical connections
Specifications

Specifications

Knife Switch
3 in.
Plastic
1 in.
Single pole, Single throw
45.35 g
3.81 cm
Electrical connections
7.62 cm
For good electrical connections.
2.54 cm
0.1 lb.
1.5 in.
Videos
SDS
Documents

Documents

Promotions

Promotions