Koehler™ Instrument Copper-Strip Test Bomb
Precision machined stainless steel bomb inserts in copper corrosion bath for testing aviation fuels and natural gasoline
Supplier:  Koehler™ Instrument K25200
Catalog No. KL-K25200
Explore available promotions
Description
Description
  • Withstands test pressure of 100psi (689kPa) per specifications
  • Threaded cap with O-ring gasket and knurled circumference tightens by hand to a positive seal
  • A 1/8 groove in the bomb threads permits safe, gradual release of pressure when opening the bomb
  • Weight: 0.91kg (2 lb.)
Compliance
Conforms to ASTM D130 and related test specifications
Specifications
Specifications
Copper Strip Test Bomb
Stainless steel
Product Suggestions
Product Suggestions