Koehler™ Instrument Smoke Point Lamp and Accessories
Supplier:  Koehler™ Instrument K27010
 View more versions of this product
Catalog No. KL-K27010
Explore available promotions
Description
Description
Specifications
Specifications
Interchangeable Candle
Wick tube and air vent
Smoke point lamp
Plated Brass
Burns fuel samples under controlled conditions for smoke point determinations of aviation turbine fuels and similar products