missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Koehler™ Instrument KV1000 Digital Constant Temperature Kinematic Viscosity Bath

Constant temperature bath for kinematic viscosity testing of petroleum products

Supplier:  Koehler™ Instrument K2337600000

 View more versions of this product

Catalog No. 13-626-100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Kinematic viscosity is of primary importance in the design and selection of a wide range of petroleum products.

  • Accommodates six round 2 in. dia. (51mm) viscometer holders
  • Test temperatures of up to 150°C (302°F) can be selected
  • Temperature limit control permits operator to select an overtemperature cutoff point to protect against accidental overheating
  • Control unit includes immersion heater, circulating stirrer and temperature probe
  • Bath Medium: water or white technical oil (sold separately)
  • Thermometers sold separately

Compliance

Complies with ASTM D445, D6074, D6158; IP 71; ISO 3104; DIN 51550;FTM 791-305; and NF T 60-100 specifications.

Specifications

Specifications

22 L
KV1000
25 lb.
13.5 x 20 in. (34.6 x 50.8 cm)
12 in. (31cm)
Videos
SDS
Documents

Documents

Promotions

Promotions