DWK Life Sciences Kimble™ Kontes™ Allihn Condensers with Full Length Joints
Allihn reflux condenser with a standard taper drip joint and a standard taper outer
Supplier: DWK Life Sciences 4310002420
Description
- 24/40 standard taper joints
- £2 hose connection
Specifications
Standard taper drip joint at bottom and a standard taper outer joint at top | |
24/40 | |
37 cm | |
14.5 in. | |
Borosilicate Glass | |
0.37 in. |
7.8 in. | |
Refluxing operations | |
Allihn Condenser | |
20 cm | |
0.952 cm | |
USP Type I and ASTM E438 Type I, Class A |