Learn More
Kontes™ Membrane Filtration Glassware from Restek
Used for routine filtration of corrosive liquids and removal of particles from HPLC solvents
Supplier: Chromatographic RKKT9537810000
Description
- Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
- Support base has a coarse porosity glass frit and an integral vacuum connection, located above the drip tip that prevents contamination of the vacuum line with filtrate droplets
Specifications
| Microfiltration Apparatus | |
| Funnel (1 L) | |
| 47 mm |
| Glass | |
| 1 L |