Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Septum Sample Tube Caps
For 5mm, 7mm and 10mm NMR Tubes
$229.50 - $361.93
Specifications
Product Type Septa
Material Rubber
For Use With (Application) Provides penetration point for syringe or cannulation additions and useful as a closure or for sample outgassing
View More Specs
Products 3
Catalog Number Mfr. No. Color For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Color For Use With (Equipment) Price Quantity  
K8970980010
View Documents
DWK Life Sciences
8970980010
Red 10 mm Thin-walled NMR Tube
Case of 100 Each for $229.50
Only null left
K8970980005
View Documents
DWK Life Sciences
8970980005
Red 5 mm Thin-walled NMR Tube
Case of 100 Each for $353.20
Only null left
K8970980007
View Documents
DWK Life Sciences
8970980007
Red 7 mm Thin-walled NMR Tube
Case of 100 Each for $361.93
Only null left
Description
Description
For 5mm, 7mm and 10mm NMR Tubes
Soft, natural rubber allows easy penetration with a syringe needle
  • Dual seal, inside tube and outer sleeve
  • Ideal for thin wall tubes
  • Provides penetration point for syringe or cannulation additions
  • Useful as a closure or for sample outgassing
Specifications
Specifications
Septa
Rubber
Provides penetration point for syringe or cannulation additions and useful as a closure or for sample outgassing
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'