DWK Life Sciences Kimble™ Kontes™ PTFE Plug for Straight Bore Stopcocks, Size 2
PTFE Stopcock Plug size 11/25 for 2mm bore size
Supplier:  DWK Life Sciences 8210010002
Catalog No. K821001-0002
Explore available promotions
Description
Description
  • PTFE stopcock plug has a 1:5 taper
  • PTFE is chemically-resistant and self-lubricating
Specifications
Specifications
Stopcock Plug
Borosilicate Glass
ASTM E438
Straight bore, PTFE plug, 1:5 taper threaded pressure nut