KOVA™ Glasstic™ Slide 10 with Grids DFS Item
Use optically clear plastic for microscopic analysis with same refractive index as glass. Glasstic™ Slide 10 with Grids is a 10-chambered slide with a hemocytometer-type grid.
Supplier:  KOVA™ 87144E
Catalog No. 22-270-141
Explore available promotions
Description
Description
Use optically clear plastic for microscopic analysis with same refractive index as glass. Glasstic™ Slide 10 with Grids is a 10-chambered slide with a hemocytometer-type grid.
KOVA Glasstic Slides sets the standard for accurate, reproducible, cost-effective and time-efficient standardized microscopic analysis. KOVA Glasstic Slides are extremely time-efficient and cost-effective.
  • Manufactured from optically clear plastic with same refractive index as glass
  • Each 10-chambered Glasstic Slide features 10 individual fully segregated specimen wells
  • Capillary action reduces pipetting errors
  • Suspends sediments homogeneously throughout the specimen
  • Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
  • Chamber walls prevent specimen leakage and cross-contamination
  • Glasstic slide grid is in the focal plane of the specimen, allowing for use over a range of magification
  • Provides results that have high correlation with results obtained with precision hemocytometers: red blood cells: r: 0.99; white blood cells: r: 0.99; platelets: r: 0.93
Specifications
Specifications
Glasstic Slide 10:100 Slides with Grids
Yes
10
Microscopic examination of Urine and other body fluids
Plastic with the optical qualities of glass
Each
Product Suggestions
Product Suggestions