missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-(-)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals
$55.00 - $611.55
Chemical Identifiers
CAS | 20312-36-1 |
---|---|
Molecular Formula | C9H9O3 |
Molecular Weight (g/mol) | 165.17 |
MDL Number | MFCD00004244 |
InChI Key | VOXXWSYKYCBWHO-QMMMGPOBSA-M |
Synonym | l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid |
PubChem CID | 444718 |
ChEBI | CHEBI:43065 |
IUPAC Name | (2S)-2-hydroxy-3-phenylpropanoic acid |
SMILES | O[C@@H](CC1=CC=CC=C1)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1820303
|
Thermo Scientific Chemicals
A1820303 |
1 g |
Each for $55.00
|
|
|||||
AAA1820306
|
Thermo Scientific Chemicals
A1820306 |
5 g |
Each for $184.96
|
|
|||||
AAA1820314
|
Thermo Scientific Chemicals
A1820314 |
25 g |
Each for $611.55
|
|
|||||
Description
L-(-)-3-Phenyllactic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-(-)-3-Phenyllactic acid is used as pharmaceutical intermediate.
Solubility
Solubility in water gives very faint turbidity.Â
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
20312-36-1 | |
165.17 | |
VOXXWSYKYCBWHO-QMMMGPOBSA-M | |
444718 | |
(2S)-2-hydroxy-3-phenylpropanoic acid |
C9H9O3 | |
MFCD00004244 | |
l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid | |
CHEBI:43065 | |
O[C@@H](CC1=CC=CC=C1)C([O-])=O |
Specifications
20312-36-1 | |
C9H9O3 | |
1 g | |
l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid | |
−24° (c=1 in Water) | |
(2S)-2-hydroxy-3-phenylpropanoic acid | |
444718 | |
166.18 | |
L-(-)-3-Phenyllactic acid |
122°C to 126°C | |
MFCD00004244 | |
2209792 | |
VOXXWSYKYCBWHO-QMMMGPOBSA-M | |
O[C@@H](CC1=CC=CC=C1)C([O-])=O | |
165.17 | |
CHEBI:43065 | |
98% |
Safety and Handling
EINECSNumber : 243-726-5
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only