missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(-)-3-Phenyllactic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 150920010

 View more versions of this product

Catalog No. AC150920010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(-)-3-Phenyllactic acid
121°C to 125°C
Authentic
Glass bottle
C6H5CH2CH(OH)CO2H
1 g
-18.5
Solubility in water: soluble
O[C@@H](CC1=CC=CC=C1)C([O-])=O
165.17
CHEBI:43065
98%
20312-36-1
Beige-Brown to White
98%
C9H9O3
MFCD00004244
−18.50° (24°C c=1,CH3COOH)
l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid
VOXXWSYKYCBWHO-QMMMGPOBSA-M
(2S)-2-hydroxy-3-phenylpropanoic acid
444718
166.18
Crystals, Powder or Flakes

Chemical Identifiers

20312-36-1
165.17
VOXXWSYKYCBWHO-QMMMGPOBSA-M
444718
(2S)-2-hydroxy-3-phenylpropanoic acid
C9H9O3
MFCD00004244
l---3-phenyllactic acid, s-2-hydroxy-3-phenylpropionic acid, s-2-hydroxy-3-phenylpropanoic acid, l--3-phenyllactic acid, s---3-phenyllactic acid, l-3-phenyllactic acid, d-3-phenyllactic acid, 2s-2-hydroxy-3-phenylpropanoic acid, s-3-phenyllactic acid, s---2-hydroxy-3-phenylpropionic acid
CHEBI:43065
O[C@@H](CC1=CC=CC=C1)C([O-])=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 243-726-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only