missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Adrenaline, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0491114

 View more versions of this product

Catalog No. AAL0491114


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-Adrenaline
216°C to 219°C
MFCD00002204
UN2811
Air and light sensitive
epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin
−52° (c=1 in 0.6N HCl)
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
5816
183.2
51-43-4
C9H13NO3
25 g
2368277
14,3619
UCTWMZQNUQWSLP-VIFPVBQESA-N
CNCC(C1=CC(=C(C=C1)O)O)O
183.207
CHEBI:28918
≥98%

Chemical Identifiers

51-43-4
183.207
UCTWMZQNUQWSLP-VIFPVBQESA-N
5816
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
C9H13NO3
MFCD00002204
epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin
CHEBI:28918
CNCC(C1=CC(=C(C=C1)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P314-P330-P361-P501c

H301+H331-H310-H315-H319-H335-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 200-098-7

RTECSNumber : DO2625000

TSCA : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only