missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Alanine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1017822

 View more versions of this product

Catalog No. AAA1017822


Explore available promotions

Description

Description

L-Alanine methyl ester hydrochloride is used to make in-vivo measurement of glucose and alanine metabolism in studies of patients with diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solubility
Soluble in Water (100 mg/ml).

Notes
Hygroscopic store under inert gas Store away from strong oxidizing agents.
Specifications

Specifications

106°C to 110°C
2491-20-5
MFCD00063663
l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
IYUKFAFDFHZKPI-DOIAFFBONA-N
methyl (2S)-2-aminopropanoate;hydrochloride
139.58
+6.5° (c=10 in methanol)
Hygroscopic
L-Alanine methyl ester hydrochloride
C4H10ClNO2
3594033
Soluble in Water (100mg/ml).
Cl.COC(=O)[C@H](C)N
100 g
2733257
139.58
99%

Chemical Identifiers

2491-20-5
139.58
IYUKFAFDFHZKPI-DOIAFFBONA-N
2733257
Cl.COC(=O)[C@H](C)N
C4H10ClNO2
MFCD00063663
l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
methyl (2S)-2-aminopropanoate;hydrochloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-652-4

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only