missing translation for 'onlineSavingsMsg'
Learn More

L-Alanyl-L-leucine, 95%, Thermo Scientific Chemicals

$1,205.70 - $1,205.70

Chemical Identifiers

CAS 3303-34-2
Molecular Formula C9H18N2O3
Molecular Weight (g/mol) 202.25
MDL Number MFCD00065106
InChI Key RDIKFPRVLJLMER-WZTWBHKBNA-N
Synonym ala-leu, l-alanyl-l-leucine, n-l-alanyl-l-leucine, h-ala-leu-oh, alanyl-dl-leucine, 2s-2-2s-2-aminopropanamido-4-methylpentanoic acid, leucine, alanyl, l-ala-l-leu, l-ala-l-leu-oh, l-leucine, l-alanyl
PubChem CID 96801
ChEBI CHEBI:73770
IUPAC Name (2S)-2-[(2S)-2-aminopropanamido]-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC(=O)[C@H](C)N)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAH6504606
View Documents
Thermo Scientific Chemicals
H6504606
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3303-34-2
202.25
RDIKFPRVLJLMER-WZTWBHKBNA-N
96801
(2S)-2-[(2S)-2-aminopropanamido]-4-methylpentanoic acid
C9H18N2O3
MFCD00065106
ala-leu, l-alanyl-l-leucine, n-l-alanyl-l-leucine, h-ala-leu-oh, alanyl-dl-leucine, 2s-2-2s-2-aminopropanamido-4-methylpentanoic acid, leucine, alanyl, l-ala-l-leu, l-ala-l-leu-oh, l-leucine, l-alanyl
CHEBI:73770
CC(C)C[C@H](NC(=O)[C@H](C)N)C(O)=O

Specifications

3303-34-2
MFCD00065106
ala-leu, l-alanyl-l-leucine, n-l-alanyl-l-leucine, h-ala-leu-oh, alanyl-dl-leucine, 2s-2-2s-2-aminopropanamido-4-methylpentanoic acid, leucine, alanyl, l-ala-l-leu, l-ala-l-leu-oh, l-leucine, l-alanyl
CC(C)C[C@H](NC(=O)[C@H](C)N)C(O)=O
5 g
96801
202.25
Solid
C9H18N2O3
1726651
RDIKFPRVLJLMER-WZTWBHKBNA-N
(2S)-2-[(2S)-2-aminopropanamido]-4-methylpentanoic acid
202.25
CHEBI:73770
95%
L-Alanyl-L-leucine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-977-1

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only