missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals L-alpha-Lecithin, granular, from soybean oil

L-alpha-Lecithin, >-97% (phospholipids) min. (as acetone insoluble), C42H80NO8P, CAS Number-8002-43-5

Supplier:  Thermo Scientific Chemicals 413100010

 View more versions of this product

Catalog No. AC413100010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-α-Lecithin
8002-43-5
Tan or Yellow
Plastic bottle
MFCD00082428
15, 5483
Solubility in water: negligible
CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
750
CHEBI:86658
≥97% (phospholipids) min. (as acetone insoluble)
Granules
Source: Soybean
35mg KOH/g max.
Authentic
C42H80NO8P
1 kg
3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine
JLPULHDHAOZNQI-JLOPVYAASA-N
[(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
16213884
750
0.02% max. (in hexane)

Chemical Identifiers

8002-43-5
750
JLPULHDHAOZNQI-JLOPVYAASA-N
16213884
[(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
C42H80NO8P
MFCD00082428
3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine
CHEBI:86658
CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-307-2

RTECSNumber : OG7565000

TSCA : TSCA

storageNote1 : Air sensitive

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only