missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(-)-alpha-Methylbenzylamine, 99+%, (99% ee), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 151830250

 View more versions of this product

Catalog No. AC151830250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
Specifications

Specifications

L(-)-α-Methylbenzylamine
-10.0°C
0.9400g/mL
71°C
Authentic
C8H11N
C6H5CH(CH3)NH2
12,1093
0.94
(20°C, 589nm) < -39° (neat)
Solubility in water: 42g/L (20°C). Other solubilities: soluble in most organic solvents,miscible with alcohols and ethers
CC(C1=CC=CC=C1)N
121.18
CHEBI:35321
99+%
2627-86-3
Colorless
187.0°C to 189.0°C
> 98.5%
Glass bottle
1.5250 to 1.5270
25 g
01,838; 02,271; 03,199; 05,441; 06,457; 11,411; 12,319; 13,185; 14,257; 17,276
14,6026
s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine
RQEUFEKYXDPUSK-ZETCQYMHSA-N
(1S)-1-phenylethanamine
75818
121.18
Liquid

Chemical Identifiers

2627-86-3
121.18
s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine
CHEBI:35321
CC(C1=CC=CC=C1)N
C8H11N
RQEUFEKYXDPUSK-ZETCQYMHSA-N
75818
(1S)-1-phenylethanamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 220-098-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only