missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Arginine, 99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210073690
Specifications
L-Arginine free base | |
White | |
C6H14N4O2 | |
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l | |
NC(CCCN=C(N)N)C(O)=O | |
500 g | |
6322 | |
174.2 |
74-79-3 | |
99% | |
MFCD00002635 | |
ODKSFYDXXFIFQN-UHFFFAOYNA-N | |
2-amino-5-[(diaminomethylidene)amino]pentanoic acid | |
174.20 | |
CHEBI:16467 | |
Powder |
Chemical Identifiers
74-79-3 | |
174.20 | |
ODKSFYDXXFIFQN-UHFFFAOYNA-N | |
6322 | |
NC(CCCN=C(N)N)C(O)=O |
C6H14N4O2 | |
MFCD00002635 | |
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l | |
CHEBI:16467 |
Safety and Handling
EINECSNumber : 200-811-1
Recommended Storage : Store at room temperature.