missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Arginine, 99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210073691

 View more versions of this product

Catalog No. ICN10073691


Explore available promotions

Description

Description

Specifications

Specifications

L-Arginine free base
White
C6H14N4O2
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
NC(CCCN=C(N)N)C(O)=O
1 kg
6322
174.2
74-79-3
99%
MFCD00002635
ODKSFYDXXFIFQN-UHFFFAOYNA-N
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
174.20
CHEBI:16467
Powder

Chemical Identifiers

74-79-3
174.20
ODKSFYDXXFIFQN-UHFFFAOYNA-N
6322
NC(CCCN=C(N)N)C(O)=O
C6H14N4O2
MFCD00002635
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
CHEBI:16467
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-811-1

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions