Learn More
Thermo Scientific™ L-Arginine-HCl, 13C6 for SILAC
Study protein expression by mass spectrometry with mass variants of lysine, arginine and other amino acids to perform stable isotope labeling with amino acids in cell culture.
$1404.00 - $1404.00
Specifications
For Use With (Equipment) | Mass Spectrometer |
---|---|
Product Type | Amino Acids |
Content And Storage | Store at room temperature. |
Label or Dye | Mass Isotope of Amino Acid |
Shipping Condition | Room Temperature |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PI88210
|
Thermo Scientific™
88210 |
50 mg |
Each for $1,404.00
|
|
|||||
Description
Thermo Scientific™ Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits.
SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of “heavy“ 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.Highlights:
Efficient – 100% label incorporation into proteins of living cells
Flexible – different isotopes of heavy and/or light amino acids for arginine, lysine, leucine and proline enable the quantitation of peptides derived from MS-grade proteases
Multiplex capabilities – several alternative isotopes of arginine and lysine are available that allow the analysis of multiple treatment conditions in each experiment
High-quality supplements – heavy amino acids with >99% isotope purity
Specifications
Mass Spectrometer | |
Store at room temperature. | |
Room Temperature | |
Protein | |
Protein Labeling |
Amino Acids | |
Mass Isotope of Amino Acid | |
Cells | |
Mass Spectrometry | |
Solid |
For Research Use Only. Not for use in diagnostic procedures.