missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Ascorbic Acid 2-Phosphate Sesquimagnesium Salt Hydrate 98.0+%, TCI America™
Supplier: TCI America A25215G
Specifications
L-Ascorbic Acid 2-Phosphate Sesquimagnesium Salt Hydrate | |
C12H18Mg3O18P2 | |
5 g | |
[Mg].[Mg].[Mg].OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O.OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O | |
585.12 | |
579.07 | |
Crystalline Powder |
113170-55-1 | |
MFCD08063372 | |
IRBRZNUQJUHOBT-UHFFFAOYNA-N | |
bis({[5-(1,2-dihydroxyethyl)-4-hydroxy-2-oxo-2,5-dihydrofuran-3-yl]oxy}phosphonic acid) trimagnesium | |
129894323 | |
≥98.0% (HPLC) |
Chemical Identifiers
113170-55-1 | |
585.12 | |
IRBRZNUQJUHOBT-UHFFFAOYNA-N | |
bis({[5-(1,2-dihydroxyethyl)-4-hydroxy-2-oxo-2,5-dihydrofuran-3-yl]oxy}phosphonic acid) trimagnesium |
C12H18Mg3O18P2 | |
MFCD08063372 | |
129894323 | |
[Mg].[Mg].[Mg].OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O.OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O |
Safety and Handling
EINECSNumber : (5)-5982
TSCA : No