missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Ascorbic acid, ≥98%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210076925

 View more versions of this product

Catalog No. ICN10076925


Explore available promotions

Description

Description

Specifications

Specifications

L-Ascorbic acid
White to Yellow
≥98%
MFCD00064328
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
176.12
CHEBI:29073
Crystalline Powder
50-81-7
1.65g/cm3
C6H8O6
25 g
CIWBSHSKHKDKBQ-DOMZIZNONA-N
(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
54670067
176.1

Chemical Identifiers

50-81-7
176.12
CIWBSHSKHKDKBQ-DOMZIZNONA-N
54670067
(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
C6H8O6
MFCD00064328
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
CHEBI:29073
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-066-2

storageNote1 : Protect from light.

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions