missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-(+)-Ascorbic acid, ACS, 99+%, Thermo Scientific Chemicals

$47.56 - $314.66

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3623714
View Documents
Thermo Scientific Chemicals
03623714
25 g
Each for $47.56
 
AA3623722
View Documents
Thermo Scientific Chemicals
03623722
100 g
Each for $86.96
 
AA3623736
View Documents
Thermo Scientific Chemicals
03623736
500 g
Each for $314.66
 
Description

Description

It is all important nutrient and antioxidant. L-ascorbic acid is frequently practiced as a processing aid or as a preservative in certain nutrients or food components. It is likewise utilized in the synthetic polymer industry, in photo processing, and in metal technology. Miscellaneous uses have been proposed in cosmetics, fibers, preservation of blood, preservation of cut plants, cleaning agents, and in assay reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-81-7
176.12
CIWBSHSKHKDKBQ-DOMZIZNONA-N
54670067
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
C6H8O6
MFCD00064328
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
CHEBI:29073
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

50-81-7
1.65 g/mL
C6H8O6
25 g
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
CIWBSHSKHKDKBQ-DOMZIZNONA-N
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
54670067
176.12
Powder
∽191°C (decomposition)
>99+%
MFCD00064328
84272
Soluble in water. Insoluble in ether,chloroform,benzene,petroleum ether,oils,fats and fat solvents. L-ascorbic acid means that the compound′s stereochemistry is related to that of the levorotatory enantiomer of glyceraldehyde.
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
176.12
CHEBI:29073
ACS Reagent
L-Ascorbic acid, For analysis ACS
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-066-2

RTECSNumber : CI7650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only