missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Ascorbic Acid (Crystalline/Certified ACS), Fisher Chemical™

Not for human use

Supplier:  Thermo Fisher Scientific FLA6125

 View more versions of this product

Catalog No. A61-25

Explore available promotions

Description

Description

Specifications

Specifications

L-Ascorbic Acid
190°C
1.65g/cm³
≥99.0 %
C6H8O6
MFCD00064328
+20.5 to +21.5° (+ or -)
CIWBSHSKHKDKBQ-DOMZIZNONA-N
(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
54670067
176.13
Certified ACS
50-81-7
Yellow
0.1% max.
Glass Bottle
CH2OHCHOHCHCOH:COHCOO
25 g
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
176.12
CHEBI:29073
≥99.0%
Solid

Chemical Identifiers

50-81-7
176.12
CIWBSHSKHKDKBQ-DOMZIZNONA-N
54670067
(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
C6H8O6
MFCD00064328
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
CHEBI:29073
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions