missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L(+)-Asparagine monohydrate, specified according the requirements of Ph.Eur., Thermo Scientific™
Supplier: Thermo Scientific Chemicals 401482500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
L(+)-Asparagine monohydrate | |
C4H10N2O4 | |
+ 35.00 | |
Solubility in water: 30g/L water (20°C). Other solubilities: soluble in acids and alkalies, practically insoluble in methanol, ethanol, ether, and benzene | |
C(C(C(=O)O)N)C(=O)N.O | |
250g | |
170358 |
5794-13-8 | |
H2NCOCH2CH(NH2)CO2H·H2O | |
l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521 | |
RBMGJIZCEWRQES-DKWTVANSSA-N | |
(2S)-2,4-diamino-4-oxobutanoic acid;hydrate | |
150.134 | |
150.14 |
RUO â Research Use Only