missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Aspartic acid di-tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals
$155.22 - $586.21
Chemical Identifiers
CAS | 1791-13-5 |
---|---|
Molecular Formula | C12H24ClNO4 |
Molecular Weight (g/mol) | 281.777 |
MDL Number | MFCD00034851 |
InChI Key | GVLZIMQSYQDAHB-QRPNPIFTSA-N |
Synonym | h-asp otbu-otbu.hcl, l-aspartic acid di-tert-butyl ester hydrochloride, s-di-tert-butyl 2-aminosuccinate hydrochloride, h-asp otbu-otbu hcl, di-tert-butyl l-aspartate hydrochloride, l-aspartic acid, bis 1,1-dimethylethyl ester, hydrochloride, l-aspartic acid di-t-butyl ester hydrochloride, h-asp obut-obut·hcl, 1,4-di-tert-butyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-otbucl |
PubChem CID | 13768146 |
IUPAC Name | ditert-butyl (2S)-2-aminobutanedioate;hydrochloride |
SMILES | CC(C)(C)OC(=O)CC(C(=O)OC(C)(C)C)N.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6371606
|
Thermo Scientific Chemicals
H6371606 |
5 g |
Each for $155.22
|
|
|||||
AAH6371614
|
Thermo Scientific Chemicals
H6371614 |
25 g |
Each for $586.21
|
|
|||||
Description
L-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
1791-13-5 | |
281.777 | |
GVLZIMQSYQDAHB-QRPNPIFTSA-N | |
13768146 | |
CC(C)(C)OC(=O)CC(C(=O)OC(C)(C)C)N.Cl |
C12H24ClNO4 | |
MFCD00034851 | |
h-asp otbu-otbu.hcl, l-aspartic acid di-tert-butyl ester hydrochloride, s-di-tert-butyl 2-aminosuccinate hydrochloride, h-asp otbu-otbu hcl, di-tert-butyl l-aspartate hydrochloride, l-aspartic acid, bis 1,1-dimethylethyl ester, hydrochloride, l-aspartic acid di-t-butyl ester hydrochloride, h-asp obut-obut·hcl, 1,4-di-tert-butyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-otbucl | |
ditert-butyl (2S)-2-aminobutanedioate;hydrochloride |
Specifications
1791-13-5 | |
MFCD00034851 | |
h-asp otbu-otbu.hcl, l-aspartic acid di-tert-butyl ester hydrochloride, s-di-tert-butyl 2-aminosuccinate hydrochloride, h-asp otbu-otbu hcl, di-tert-butyl l-aspartate hydrochloride, l-aspartic acid, bis 1,1-dimethylethyl ester, hydrochloride, l-aspartic acid di-t-butyl ester hydrochloride, h-asp obut-obut·hcl, 1,4-di-tert-butyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-otbucl | |
CC(C)(C)OC(=O)CC(C(=O)OC(C)(C)C)N.Cl | |
5 g | |
13768146 | |
98% | |
L-Aspartic acid di-tert-butyl ester hydrochloride |
C12H24ClNO4 | |
5652807 | |
GVLZIMQSYQDAHB-QRPNPIFTSA-N | |
ditert-butyl (2S)-2-aminobutanedioate;hydrochloride | |
281.777 | |
281.78 | |
Solid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only