missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid monosodium salt monohydrate, 97%

CAS: 3792-50-5 | C4H11NNaO6 | 192.123 g/mol

$48.69 - $130.12

Chemical Identifiers

CAS 3792-50-5
Molecular Formula C4H11NNaO6
Molecular Weight (g/mol) 192.123
InChI Key VJDUMAWJHIHDHJ-SQGDDOFFSA-N
Synonym monosodium l-aspartate dihydrate
PubChem CID 129893661
IUPAC Name (2S)-2-aminobutanedioic acid;sodium;dihydrate
SMILES C(C(C(=O)O)N)C(=O)O.O.O.[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC451620250
SDS View Documents
Thermo Scientific Chemicals
451620250
25 g Plastic Bottle
Each for $48.69
Only null left
 
AC451621000
SDS View Documents
Thermo Scientific Chemicals
451621000
100 g Plastic Bottle
Each for $130.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3792-50-5
192.123
monosodium l-aspartate dihydrate
(2S)-2-aminobutanedioic acid;sodium;dihydrate
C4H11NNaO6
VJDUMAWJHIHDHJ-SQGDDOFFSA-N
129893661
C(C(C(=O)O)N)C(=O)O.O.O.[Na]

Specifications

3792-50-5
C4H11NNaO6
monosodium l-aspartate dihydrate
C(C(C(=O)O)N)C(=O)O.O.O.[Na]
25 g
129893661
97%
97%
Plastic Bottle
VJDUMAWJHIHDHJ-SQGDDOFFSA-N
(2S)-2-aminobutanedioic acid;sodium;dihydrate
192.123
173.1
L-Aspartic acid monosodium salt monohydrate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only