missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-beta-Homoleucine hydrochloride, 95%, Thermo Scientific Chemicals

$257.49 - $719.52

Chemical Identifiers

CAS 96386-92-4
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD01862871
InChI Key NXVYPYHWONGEFQ-RGMNGODLSA-N
Synonym l-beta-homoleucine hydrochloride, a-holeu-oh.hcl, s-3-amino-5-methylhexanoic acid hydrochloride, 3s-3-amino-5-methylhexanoic acid hydrochloride, 3s-3-amino-5-methylhexanoic acid hydrochloride, hexanoic acid, 3-amino-5-methyl-, hydrochloride, 3s, l-beta-homoleucine-hcl, l-beta-homoleucine-hcl, l-beta-homoleucinehcl, h-, h-
PubChem CID 2761524
IUPAC Name (3S)-3-amino-5-methylhexanoic acid hydrochloride
SMILES Cl.CC(C)C[C@H](N)CC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65857MD
View Documents
Thermo Scientific Chemicals
H65857MD
250 mg
Each for $257.49
 
AAH6585703
View Documents
Thermo Scientific Chemicals
H6585703
1 g
Each for $719.52
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

96386-92-4
181.66
NXVYPYHWONGEFQ-RGMNGODLSA-N
2761524
Cl.CC(C)C[C@H](N)CC(O)=O
C7H16ClNO2
MFCD01862871
l-beta-homoleucine hydrochloride, a-holeu-oh.hcl, s-3-amino-5-methylhexanoic acid hydrochloride, 3s-3-amino-5-methylhexanoic acid hydrochloride, 3s-3-amino-5-methylhexanoic acid hydrochloride, hexanoic acid, 3-amino-5-methyl-, hydrochloride, 3s, l-beta-homoleucine-hcl, l-beta-homoleucine-hcl, l-beta-homoleucinehcl, h-, h-
(3S)-3-amino-5-methylhexanoic acid hydrochloride

Specifications

96386-92-4
C7H16ClNO2
8073252
NXVYPYHWONGEFQ-RGMNGODLSA-N
(3S)-3-amino-5-methylhexanoic acid hydrochloride
181.66
181.66
Powder
Brown to White
MFCD01862871
l-beta-homoleucine hydrochloride, a-holeu-oh.hcl, s-3-amino-5-methylhexanoic acid hydrochloride, 3s-3-amino-5-methylhexanoic acid hydrochloride, 3s-3-amino-5-methylhexanoic acid hydrochloride, hexanoic acid, 3-amino-5-methyl-, hydrochloride, 3s, l-beta-homoleucine-hcl, l-beta-homoleucine-hcl, l-beta-homoleucinehcl, h-, h-
Cl.CC(C)C[C@H](N)CC(O)=O
250 mg
2761524
95%
L-beta-Homoleucine hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only