Learn More
L-Buthionine-(S,R)-sulfoximine, 99%, Thermo Scientific Chemicals
$566.01 - $1798.26
Chemical Identifiers
CAS | 83730-53-4 |
---|---|
Molecular Formula | C8H18N2O3S |
Molecular Weight (g/mol) | 222.31 |
MDL Number | MFCD00067000 |
InChI Key | KJQFBVYMGADDTQ-CVSPRKDYSA-N |
Synonym | l-buthionine-sulfoximine, l-buthionine sulfoximine, 2s-2-amino-4-butylsulfonimidoyl butanoic acid, l-buthionine-s,r-sulfoximine, l-buthionine s,r-sulfoximine, l-bso, butanoic acid, 2-amino-4-s-butylsulfonimidoyl-, 2s, l-buthionine sr-sulfoximine, buthionine-s,r-sulfoximine, l, l-buthionine-s,r-sulphoximine |
PubChem CID | 119565 |
IUPAC Name | (2S)-2-amino-4-(butylsulfonimidoyl)butanoic acid |
SMILES | CCCCS(=N)(=O)CCC(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC235520010
|
Thermo Scientific Chemicals
235520010 |
1 g | Glass bottle |
Each for $566.01
|
|
||||
AC235520050
|
Thermo Scientific Chemicals
235520050 |
5 g | Glass bottle |
Each for $1,798.26
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
83730-53-4 | |
222.31 | |
KJQFBVYMGADDTQ-CVSPRKDYSA-N | |
119565 | |
CCCCS(=N)(=O)CCC(C(=O)O)N |
C8H18N2O3S | |
MFCD00067000 | |
l-buthionine-sulfoximine, l-buthionine sulfoximine, 2s-2-amino-4-butylsulfonimidoyl butanoic acid, l-buthionine-s,r-sulfoximine, l-buthionine s,r-sulfoximine, l-bso, butanoic acid, 2-amino-4-s-butylsulfonimidoyl-, 2s, l-buthionine sr-sulfoximine, buthionine-s,r-sulfoximine, l, l-buthionine-s,r-sulphoximine | |
(2S)-2-amino-4-(butylsulfonimidoyl)butanoic acid |
Specifications
83730-53-4 | |
White | |
99% | |
C8H18N2O3S | |
MFCD00067000 | |
+ 33.00 | |
Solubility in water: soluble | |
CCCCS(=N)(=O)CCC(C(=O)O)N | |
222.31 | |
222.31 | |
Fine Powder |
234.0°C to 235.0°C | |
Authentic | |
Glass bottle | |
HOOCCH(NH2)CH2CH2S(O)(=NH)(CH2)3CH3 | |
1 g | |
l-buthionine-sulfoximine, l-buthionine sulfoximine, 2s-2-amino-4-butylsulfonimidoyl butanoic acid, l-buthionine-s,r-sulfoximine, l-buthionine s,r-sulfoximine, l-bso, butanoic acid, 2-amino-4-s-butylsulfonimidoyl-, 2s, l-buthionine sr-sulfoximine, buthionine-s,r-sulfoximine, l, l-buthionine-s,r-sulphoximine | |
KJQFBVYMGADDTQ-CVSPRKDYSA-N | |
(2S)-2-amino-4-(butylsulfonimidoyl)butanoic acid | |
119565 | |
99% | |
L-Buthionine-(S, R)-sulfoximine |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
RUO – Research Use Only