missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(-)-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 263640250

 View more versions of this product

Catalog No. AC263640250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(-)-Camphorsulfonic acid
198.0°C
Authentic
Glass bottle
MFCD00150753,MFCD00064158
11,316
− 22.00
Solubility in water: soluble. Other solubilities: slightly soluble in glacial acetic acid and ethyl,acetate,practically insoluble in ether,soluble in chloroform and alcohol
CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2
232.29
CHEBI:55401
98%
35963-20-3
White to Beige
98%
C10H16O4S
25 g
16,58
l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid
MIOPJNTWMNEORI-MHPPCMCBSA-N
[(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid
5771688
232.3
Crystalline Powder

Chemical Identifiers

35963-20-3
232.29
MIOPJNTWMNEORI-MHPPCMCBSA-N
5771688
CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2
C10H16O4S
MFCD00150753,MFCD00064158
l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid
CHEBI:55401
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 252-817-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only