Learn More
L(-)-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 263640050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
L(-)-Camphorsulfonic acid | |
198.0°C | |
Authentic | |
Glass bottle | |
MFCD00150753,MFCD00064158 | |
11,316 | |
− 22.00 | |
Solubility in water: soluble. Other solubilities: slightly soluble in glacial acetic acid and ethyl,acetate,practically insoluble in ether,soluble in chloroform and alcohol | |
CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2 | |
232.29 | |
CHEBI:55401 | |
98% |
35963-20-3 | |
White to Beige | |
98% | |
C10H16O4S | |
5 g | |
16,58 | |
l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid | |
MIOPJNTWMNEORI-MHPPCMCBSA-N | |
[(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid | |
5771688 | |
232.3 | |
Crystalline Powder |
Chemical Identifiers
35963-20-3 | |
232.29 | |
MIOPJNTWMNEORI-MHPPCMCBSA-N | |
5771688 | |
CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2 |
C10H16O4S | |
MFCD00150753,MFCD00064158 | |
l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid | |
CHEBI:55401 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 252-817-9
RUO – Research Use Only