missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Carnitine Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America C305825G

 View more versions of this product

Catalog No. C305825G


Explore available promotions

Description

Description

Specifications

Specifications

L-Carnitine Hydrochloride
139°C
C7H16ClNO3
25 g
JXXCENBLGFBQJM-FYZOBXCZSA-N
[(2R)-3-carboxy-2-hydroxypropyl]trimethylazanium chloride
656657
≥98.0% (T)
6645-46-1
White
MFCD00066100
r-3-carboxy-2-hydroxy-n,n,n-trimethylpropan-1-aminium chloride, l-carnitine hcl, unii-j3y5e6iks3, l-carnitine hydrochloride, levocarnitine chloride, j3y5e6iks3, r-carnitine hydrochloride, carnitine l-form hydrochloride, l-3-carboxy-2-hydroxypropyl trimethylammonium chloride
[Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O
197.66
197.66
Crystalline Powder

Chemical Identifiers

6645-46-1
197.66
JXXCENBLGFBQJM-FYZOBXCZSA-N
656657
[Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O
C7H16ClNO3
MFCD00066100
r-3-carboxy-2-hydroxy-n,n,n-trimethylpropan-1-aminium chloride, l-carnitine hcl, unii-j3y5e6iks3, l-carnitine hydrochloride, levocarnitine chloride, j3y5e6iks3, r-carnitine hydrochloride, carnitine l-form hydrochloride, l-3-carboxy-2-hydroxypropyl trimethylammonium chloride
[(2R)-3-carboxy-2-hydroxypropyl]trimethylazanium chloride
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0253

RTECSNumber : BP2979100

TSCA : No

SDS
Documents

Documents

Promotions

Promotions