missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(+)-Citrulline, 99%, Thermo Scientific Chemicals

$230.43 - $230.43

Chemical Identifiers

CAS 372-75-8
Molecular Formula C6H13N3O3
Molecular Weight (g/mol) 175.19
MDL Number MFCD00064397
InChI Key RHGKLRLOHDJJDR-UHFFFAOYNA-N
Synonym l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l
PubChem CID 9750
ChEBI CHEBI:16349
IUPAC Name (2S)-2-amino-5-(carbamoylamino)pentanoic acid
SMILES NC(CCCNC(N)=O)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC110470250
SDS View Documents
Thermo Scientific Chemicals
110470250
25 g Glass bottle
Each for $230.43
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

372-75-8
175.19
RHGKLRLOHDJJDR-UHFFFAOYNA-N
9750
(2S)-2-amino-5-(carbamoylamino)pentanoic acid
C6H13N3O3
MFCD00064397
l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l
CHEBI:16349
NC(CCCNC(N)=O)C(O)=O

Specifications

372-75-8
White
0.3% max. (1 g, 105°C, 3 hrs)
99%
C6H13N3O3
MFCD00064397
15, 2330
l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l
RHGKLRLOHDJJDR-UHFFFAOYNA-N
(2S)-2-amino-5-(carbamoylamino)pentanoic acid
175.19
CHEBI:16349
99%
L(+)-Citrulline
214°C
0.1% max.
Authentic
Glass bottle
H2NCONH(CH2)3CH(NH2)CO2H
25 g
+ 25.5
Solubility in water: 200g/L (20°C). Other solubilities: insoluble in methanol and ethanol
NC(CCCNC(N)=O)C(O)=O
95.0% min. (c=10, H2O, 430nm) 1cm cell
9750
175.19
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-759-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only