missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Cysteine, 99.5%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210144490

 View more versions of this product

Catalog No. ICN10144490


Explore available promotions

Description

Description

Specifications

Specifications

L-Cysteine
White
C3H7NO2S
XUJNEKJLAYXESH-REOHCLBHSA-N
(2R)-2-amino-3-sulfanylpropanoic acid
121.154
CHEBI:17561
Crystalline Powder
52-90-4
99.5%
l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
C(C(C(=O)O)N)S
500 g
5862
121.16

Chemical Identifiers

52-90-4
121.154
l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
CHEBI:17561
C(C(C(=O)O)N)S
C3H7NO2S
XUJNEKJLAYXESH-REOHCLBHSA-N
5862
(2R)-2-amino-3-sulfanylpropanoic acid
Videos
SDS
Documents

Documents

Promotions

Promotions