missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Cysteine Hydrochloride, Monohydrate, Multi-Compendial, U.S.P., J.T. Baker™

$1538.24 - $1538.24

Chemical Identifiers

CAS 7048-04-6
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
Synonym l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
PubChem CID 23462
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
02003183
View Documents
Avantor
207105
100 g Poly Bottle
Each for $1,538.24
 
Description

Description

Specifications

Chemical Identifiers

7048-04-6
175.63
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
(2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
C3H10ClNO3S
QIJRTFXNRTXDIP-JIZZDEOASA-N
23462
C(C(C(=O)O)N)S.O.Cl

Specifications

7048-04-6
Poly Bottle
QIJRTFXNRTXDIP-JIZZDEOASA-N
(2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
23462
Multi-Compendial/USP
1L = 1.50kg
C3H10ClNO3S
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
C(C(C(=O)O)N)S.O.Cl
175.63
175.64
100 g
L-Cysteine Hydrochloride, Monohydrate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'