missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

L-Cysteine Hydrochloride Monohydrate (White Crystals), Fisher BioReagents
Supplier: Fisher BioReagents FLBP376100
Description
This amino acid is suitable for use in tissue culture systems requiring additives.Specifications
175°C | |
0.02% max. | |
1ppm max. | |
White | |
10ppm max. | |
8.5 to 12.0% | |
Not Detected | |
Glass Bottle | |
5.5 to 7.0° (+ or -) | |
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl | |
QIJRTFXNRTXDIP-JIZZDEOASA-N | |
(2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride | |
100 g | |
175.64 | |
Solid |
L-Cysteine Hydrochloride Monohydrate | |
7048-04-6 | |
1 to 2 | |
19.89 to 20.29% | |
Pass Test | |
98.5 to 101 % | |
C3H10ClNO3S | |
0.1% max. | |
98.0% min. | |
Cysteine Hydrochloride Monohydrate | |
C(C(C(=O)O)N)S.O.Cl | |
175.63 | |
23462 | |
98.5 to 101% |
Chemical Identifiers
7048-04-6 | |
175.63 | |
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl | |
(2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride |
C3H10ClNO3S | |
QIJRTFXNRTXDIP-JIZZDEOASA-N | |
23462 | |
C(C(C(=O)O)N)S.O.Cl |
Safety and Handling

Recommended Storage : RT