Learn More
L-Dihydroorotic acid, 98%, Thermo Scientific Chemicals
$277.19 - $1089.55
Chemical Identifiers
CAS | 5988-19-2 |
---|---|
Molecular Formula | C5H6N2O4 |
Molecular Weight (g/mol) | 158.113 |
MDL Number | MFCD00085339 |
InChI Key | UFIVEPVSAGBUSI-REOHCLBHSA-N |
Synonym | l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate |
PubChem CID | 439216 |
ChEBI | CHEBI:17025 |
IUPAC Name | (4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid |
SMILES | C1C(NC(=O)NC1=O)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6042903
|
Thermo Scientific Chemicals
H6042903 |
1 g |
Each for $277.19
|
|
|||||
AAH6042906
|
Thermo Scientific Chemicals
H6042906 |
5 g |
Each for $1,089.55
|
|
|||||
Description
L-Dihydroorotic acid a metabolite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Dihydroorotic acid a metabolite.
Solubility
Soluble in water (partly), and dimethyl formamide.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
5988-19-2 | |
158.113 | |
UFIVEPVSAGBUSI-REOHCLBHSA-N | |
439216 | |
(4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid |
C5H6N2O4 | |
MFCD00085339 | |
l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate | |
CHEBI:17025 | |
C1C(NC(=O)NC1=O)C(=O)O |
Specifications
5988-19-2 | |
C5H6N2O4 | |
1 g | |
Soluble in water (partly),and dimethyl formamide. | |
C1C(NC(=O)NC1=O)C(=O)O | |
158.113 | |
CHEBI:17025 | |
98% |
254°C to 255°C (decomposition) | |
MFCD00085339 | |
l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate | |
UFIVEPVSAGBUSI-REOHCLBHSA-N | |
(4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid | |
439216 | |
158.11 | |
L-Dihydroorotic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only